Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLLT6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582128
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582128 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582128 Supplier Invitrogen™ Supplier No. PA582128
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSAPPSPSAP EPPKADLFEQ KVVFSGFGPI MRFSTTTSSS GRARAPSPGD YKSPHVTGSG ASAGTHKRMP ALSATPVPAD ETPETGLKEK KHKASKRSRH GPGRPKGSRN KEGTGGPAAP SLPSAQLAGF TATAASPFSG GSLVSS Highest antigen sequence indentity to the following orthologs: Rat - 87%, Mouse - 87%.

MLLT6 gene ontology annotations related to this gene include negative regulation of histone H3-K79 methylation; negative regulation of urine volume; positive regulation of sodium ion transport; positive regulation of transcription by RNA polymerase II; renal potassium excretion; renal sodium excretion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLLT6
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MLLT6
Gene Accession No. P55198
Gene Alias Af17; AI315037; ALL1-fused gene from chromosome 17 protein; MLLT6; MLLT6, PHD finger domain containing; myeloid/lymphoid or mixed lineage-leukemia translocation to 6 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocated to, 6; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 6; Myeloid/lymphoid or mixed-lineage leukemia, translocated to, 6; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 6; protein AF-17; trithorax homolog
Gene Symbols MLLT6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MLLT6. Recombinant protein control fragment (Product #RP-102310).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4302
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.