Learn More
MLN64 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 202921AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-2813
Only null left
Add to Cart
Description
Description
This gene encodes a member of a subfamily of lipid trafficking proteins that are characterized by a C-terminal steroidogenic acute regulatory domain and an N-terminal metastatic lymph node 64 domain. The encoded protein localizes to the membranes of late endosomes and may be involved in exporting cholesterol. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
MLN64
Polyclonal
Unconjugated
STARD3
CAB1, MLN 64, MLN64, Protein CAB1, STARD3
Rabbit
Antigen Affinity Chromatography
RUO
10948, 363675, 59045
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q14849, Q61542
Stard3
MLN64 Fusion Protein Ag14061
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.