Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLPH Monoclonal Antibody (OTI6E8), TrueMAB™, OriGene
SDP

Catalog No. 501682172
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2172 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2172 Supplier OriGene Supplier No. TA808503
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rab effector protein involved in melanosome transport. Serves as link between melanosome-bound RAB27A and the motor protein MYO5A.

Specifications

Antigen MLPH
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6E8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MLPH
Gene Accession No. Q9BV36
Gene Alias SLAC2-A
Gene Symbols MLPH
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 295-600 of human MLPH produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79083
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.