Learn More
Invitrogen™ MLPH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109965
Description
Immunogen sequence: EQLPLQYLAD VDTSDEESIR AHVMASHHSK RRGRASSESQ IFELNKHISA VECLLTYLEN TVVPPLAKGL GAGVRTEADV EEEALRRKLE ELTSNVSDQE TSSEEEEAKD EKAEPNRDKS VGPLPQADPE VGTAAHQTNR QEKSPQDPGD PVQYNRTTDE ELSELEDRVA VTASEVQQAE SEVSDIESRI AALRAAGLTV KPSGKPRRKS NLPIFLPRVA GKLGKRPEDP NADPSSEAKA MAVPYLLRRK FSNSLKSQGK DDDSFDRKSV YRGSLTQRNP NARKGMASHT FAKPVVAHQS.
Slac2-a (for synaptotagmin-like protein (Slp) homolog lacking C2 domains-a) links Rab27A on melanosomes with Myosin VA in melanocytes. Slac2-a binds Myosin VA through a C-terminal region and GTP-bound Rab27A through its synaptotagmin-like protein homology domain (SHD), which is located near the N-terminus. The transport of pigment and cytotoxic granules in melanocytes requires the stable formation of this complex, and thus mutations in the binding domains of the three protein components may cause albinism and/or severe immune disorders.
Specifications
MLPH | |
Polyclonal | |
Unconjugated | |
Mlph | |
2210418F23Rik; 5031433I09Rik; AW228792; D1Wsu84e; Exophilin-3; l(1)-3Rk; l1Rk3; leaden; leaden protein; Ln; Melanophilin; MGC2771; MGC59733; Mlph; OTTMUSP00000025451; SLAC2A; Slac-2a; SlaC2-a; Slp homolog lacking C2 domains a; synaptotagmin-like protein 2a | |
Rabbit | |
Affinity Chromatography | |
RUO | |
171531, 316620, 79083 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q91V27, Q9BV36 | |
Mlph | |
Recombinant fusion protein containing a sequence corresponding to amino acids 301-600 of human MLPH (NP_077006.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.