Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLSTD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543487
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543487 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543487 Supplier Invitrogen™ Supplier No. PA543487
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LRSCPKVNSV YVLVRQKAGQ TPQERVEEVL SGKLFDRLRD ENPDFREKII Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

MLSTD2 catalyzes the reduction of saturated fatty acyl-CoA with chain length C16 or C18 to fatty alcohols.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLSTD2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAR1
Gene Accession No. Q8WVX9
Gene Alias 2600011M19Rik; 2900034E22Rik; 3732409C05Rik; AI850429; FAR1; fatty acyl CoA reductase 1; fatty acyl-CoA reductase 1; male sterility domain containing 2; male sterility domain-containing protein 2; MLSTD2; PFCRD; SDR10E1; short chain dehydrogenase/reductase family 10E, member 1; UNQ2423/PRO4981
Gene Symbols FAR1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MLSTD2 (aa 31-80).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84188
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.