Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLX Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP146146
Description
MLX Polyclonal specifically detects MLX in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
MLX | |
Polyclonal | |
Immunohistochemistry 2.5 - 3.8 ug/ml, Immunohistochemistry-Paraffin 2.5 - 3.8 ug/ml, Peptide ELISA Detection limit 1:128000 | |
NP_937848.1 | |
MLX | |
Peptide with sequence C-QIVKAHQDNPHEGED corresponding to internal region according to NP_937847.1, NP_733752.1. | |
0.1 mg | |
Primary | |
This antibody is expected to recognize all three reported isoforms (NP_937848.1; NP_937847.1; NP_733752.1). | |
Store at -20C. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
BHLHD13, bHLHd13Max-like bHLHZip protein, BigMax protein, Class D basic helix-loop-helix protein 13, MAD7, MAX-like protein X, MXD7, TCFL4Protein BigMax, transcription factor-like 4, Transcription factor-like protein 4 | |
Goat | |
Affinity Purified | |
RUO | |
6945 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction