Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLX Antibody (PCRP-MLX-1G8) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308241
Description
MLX Monoclonal specifically detects MLX in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
MLX | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1-2 ug/mL, Flow Cytometry : 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence : 1-2 ug/mL, Protein Array | |
BHLHD13, bHLHd13Max-like bHLHZip protein, BigMax protein, Class D basic helix-loop-helix protein 13, MAD7, MAX-like protein X, MXD7, TCFL4Protein BigMax, transcription factor-like 4, Transcription factor-like protein 4 | |
Recombinant full-length human MLX protein | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array | |
PCRP-MLX-1G8 | |
Unconjugated | |
10mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
6945 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction