Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MLX Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562677
Description
Immunogen sequence: EEVSTLRKDV TALKIMKVNY EQIVKAHQDN PHEGEDQVSD QVKFNVFQGI MDSLFQSFNA SISVASFQEL SA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The product of this gene belongs to the family of basic helix-loop-helix leucine zipper (bHLH-Zip) transcription factors. These factors form heterodimers with Mad proteins and play a role in proliferation, determination and differentiation. This gene product may act to diversify Mad family function by its restricted association with a subset of the Mad family of transcriptional repressors, namely, Mad1 and Mad4. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
Specifications
MLX | |
Polyclonal | |
Unconjugated | |
Mlx | |
bHLHd13; BigMax alpha; BigMax protein; class D basic helix-loop-helix protein 13; hypothetical protein LOC508531; im:7146242; MAD7; MAX dimerization protein MLX; MAX-like bHLHZIP protein; max-like factor protein; max-like protein X; Mlx; MLX, MAX dimerization protein; MXD7; Protein BigMax; TCFL4; TF4; transcription factor like 4; transcription factor-like 4; transcription factor-like protein 4; wu:fb61a01; wu:fb99c01; zgc:112409 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6945 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UH92 | |
Mlx | |
Recombinant protein corresponding to Human MLX. Recombinant protein control fragment (Product #RP-103135). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction