Learn More
Invitrogen™ MLX Polyclonal Antibody

Description
Immunogen sequence: RARARRGAGR RGASLLSPKS PTLSVPRGCR EDSSHPACAK VEYAYSDNSL DPGLFVESTR KGSVVSRANS IGSTSASSV Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 91%.
Specifications
Specifications
| Antigen | MLX |
| Applications | ChIP Assay, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Mlx |
| Gene Accession No. | Q9UH92 |
| Gene Alias | bHLHd13; BigMax alpha; BigMax protein; class D basic helix-loop-helix protein 13; hypothetical protein LOC508531; im:7146242; MAD7; MAX dimerization protein MLX; MAX-like bHLHZIP protein; max-like factor protein; max-like protein X; Mlx; MLX, MAX dimerization protein; MXD7; Protein BigMax; TCFL4; TF4; transcription factor like 4; transcription factor-like 4; transcription factor-like protein 4; wu:fb61a01; wu:fb99c01; zgc:112409 |
| Gene Symbols | Mlx |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.