Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118837
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118837 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5118837 Supplier Invitrogen™ Supplier No. PA5118837
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Mlx.

The product of this gene belongs to the family of basic helix-loop-helix leucine zipper (bHLH-Zip) transcription factors. These factors form heterodimers with Mad proteins and play a role in proliferation, determination and differentiation. This gene product may act to diversify Mad family function by its restricted association with a subset of the Mad family of transcriptional repressors, namely, Mad1 and Mad4. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLX
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Mlx
Gene Accession No. O08609, Q9UH92
Gene Alias bHLHd13; BigMax alpha; BigMax protein; class D basic helix-loop-helix protein 13; hypothetical protein LOC508531; im:7146242; MAD7; MAX dimerization protein MLX; MAX-like bHLHZIP protein; max-like factor protein; max-like protein X; Mlx; MLX, MAX dimerization protein; MXD7; Protein BigMax; TCFL4; TF4; transcription factor like 4; transcription factor-like 4; transcription factor-like protein 4; wu:fb61a01; wu:fb99c01; zgc:112409
Gene Symbols Mlx
Host Species Rabbit
Immunogen A synthesized peptide derived from human MLX(Accession Q9UH92), corresponding to amino acid residues D116-I166.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21428, 360631, 6945
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.