Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLXIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106363
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106363 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106363 Supplier Invitrogen™ Supplier No. PA5106363
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MLXIP.

This gene encodes a protein that functions as part of a heterodimer to activate transcription. The encoded protein forms a heterodimer with Max-like protein X (MLX) and is involved in the regulation of genes in response to cellular glucose levels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLXIP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MLXIP
Gene Accession No. Q2VPU4, Q9HAP2
Gene Alias AW228700; bHLHe36; class E basic helix-loop-helix protein 36; KIAA0867; Mir; MLX interacting protein; Mlx interactor; MLX-interacting protein; Mlxip; MONDOA; RGD1560774; transcriptional activator MondoA
Gene Symbols MLXIP
Host Species Rabbit
Immunogen A synthesized peptide derived from human MLXIP(Accession Q9HAP2), corresponding to amino acid residues L120-W170.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 208104, 22877, 304479
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.