Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MLXIP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109966
Description
Immunogen sequence: ARAHASAAPP PPRAGPGREE PPRRQQIIHS GHFMVSSPHR EHPPKKGYDF DTVNKQTCQT YSFGKTSSCH LSIDASLTKL FECMTLAYSG KLVSPKWKNF K.
This gene encodes a protein that functions as part of a heterodimer to activate transcription. The encoded protein forms a heterodimer with Max-like protein X (MLX) and is involved in the regulation of genes in response to cellular glucose levels.
Specifications
| MLXIP | |
| Polyclonal | |
| Unconjugated | |
| MLXIP | |
| AW228700; bHLHe36; class E basic helix-loop-helix protein 36; KIAA0867; Mir; MLX interacting protein; Mlx interactor; MLX-interacting protein; Mlxip; MONDOA; RGD1560774; transcriptional activator MondoA | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22877, 304479 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 4.12 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9HAP2 | |
| MLXIP | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human MLXIP (NP_055753.3). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction