Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMAA Antibody (OTI3G5), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272726AF532
Description
MMAA Monoclonal specifically detects MMAA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
MMAA | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
MMAA | |
Full length recombinant protein of human MMAA (NP_785454) produced in HEK293T cells. | |
0.1 mL | |
166785.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3G5 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
cblA, EC 3.6, methylmalonic aciduria (cobalamin deficiency) cblA type, methylmalonic aciduria (cobalamin deficiency) type A, MGC120011, MGC120012, MGC120013, mitochondrial | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction