Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMAA Monoclonal Antibody (OTI3A3)
GREENER_CHOICE

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525385

Catalog No. PIMA525385


Only null left
Add to Cart

Description

Description

MMAA Monoclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene is involved in the translocation of cobalamin into the mitochondrion, where it is used in the final steps of adenosylcobalamin synthesis. Adenosylcobalamin is a coenzyme required for the activity of methylmalonyl-CoA mutase. Defects in this gene are a cause of methylmalonic aciduria.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MMAA
Monoclonal
0.75 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8IVH4
MMAA
Full length human recombinant protein of MMAA produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI3A3
Unconjugated
MMAA
2810018E08Rik; AI840684; cblA; methylmalonic aciduria (cobalamin deficiency) cblA type; methylmalonic aciduria (cobalamin deficiency) type A; methylmalonic aciduria type A homolog, mitochondrial; Methylmalonic aciduria type A protein, mitochondrial; MMAA; mutant adenosylcobalamin
Mouse
Affinity chromatography
RUO
166785
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.