Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMAA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114221
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114221 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114221 Supplier Invitrogen™ Supplier No. PA5114221
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GQRACLAEAI TLVESTHSRK KELAQVLLQK VLLYHREQEQ SNKGKPLAFR For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 85%; Dog: 86%; Guinea Pig: 92%; Horse: 86%; Human: 100%; Rabbit: 93%; Rat: 92%.

The protein encoded by this gene is involved in the translocation of cobalamin into the mitochondrion, where it is used in the final steps of adenosylcobalamin synthesis. Adenosylcobalamin is a coenzyme required for the activity of methylmalonyl-CoA mutase. Defects in this gene are a cause of methylmalonic aciduria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMAA
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MMAA
Gene Accession No. Q8IVH4
Gene Alias 2810018E08Rik; AI840684; cblA; methylmalonic aciduria (cobalamin deficiency) cblA type; methylmalonic aciduria (cobalamin deficiency) type A; methylmalonic aciduria type A homolog, mitochondrial; Methylmalonic aciduria type A protein, mitochondrial; MMAA; mutant adenosylcobalamin
Gene Symbols MMAA
Host Species Rabbit
Immunogen Immunogen sequence: GQRACLAEAI TLVESTHSRK KELAQVLLQK VLLYHREQEQ SNKGKPLAFR (aa 96-145).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166785
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.