Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMAA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140091
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140091 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140091 Supplier Invitrogen™ Supplier No. PA5140091
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ERGHKLSVLA VDPSSCTSGG SLLGDKTRMT ELSRDMNAYI RPSPTRGTLG GVTRTTNEAI LLCEGAGYDI ILIETVGVGQ SEFAVAD Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 93%.

The protein encoded by this gene is involved in the translocation of cobalamin into the mitochondrion, where it is used in the final steps of adenosylcobalamin synthesis. Adenosylcobalamin is a coenzyme required for the activity of methylmalonyl-CoA mutase. Defects in this gene are a cause of methylmalonic aciduria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMAA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MMAA
Gene Accession No. Q8IVH4
Gene Alias 2810018E08Rik; AI840684; cblA; methylmalonic aciduria (cobalamin deficiency) cblA type; methylmalonic aciduria (cobalamin deficiency) type A; methylmalonic aciduria type A homolog, mitochondrial; Methylmalonic aciduria type A protein, mitochondrial; MMAA; mutant adenosylcobalamin
Gene Symbols MMAA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MMAA. Recombinant protein control fragment (Product #RP-108955).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166785
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.