Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMAB Antibody (OTI2G5), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272730JF646
Description
MMAB Monoclonal specifically detects MMAB in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
MMAB | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MMAB | |
Full length recombinant protein of human MMAB (NP_443077) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI2G5 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
aquocob(I)alamin vitamin B12s adenosyltransferase, ATP:cob(I)alamin adenosyltransferase, ATP:corrinoid adenosyltransferase, ATR, cblB, c-diamide adenosyltransferase, mitochondrial, cob, Cob(I)alamin adenosyltransferase, cob(I)yrinic acid a, EC 2.5.1.17, methylmalonic aciduria (cobalamin deficiency) cblB type, methylmalonic aciduria (cobalamin deficiency) type B, Methylmalonic aciduria type B protein, MGC20496 | |
Mouse | |
Affinity Purified | |
Proteases & Other Enzymes | |
326625.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction