Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMAB Monoclonal Antibody (OTI4D12), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502166S


Description
MMAB Monoclonal antibody specifically detects MMAB in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
MMAB | |
Monoclonal | |
0.91 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9UIM3 | |
Mouse | |
Affinity chromatography | |
RUO | |
63943 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI4D12 | |
Unconjugated | |
Fkbpl | |
9130222L19Rik; aquocob(I)alamin vitamin B12s adenosyltransferase; ATP:Cob(I)alamin Adenosyltransferase; ATP:corrinoid adenosyltransferase; ATR; cblB; CFAP23; cilia and flagella associated protein 23; cob; cob(I)alamin adenosyltransferase; cob(I)yrinic acid a,c-diamide adenosyltransferase, mitochondrial; Cob(II)alamin adenosyltransferase; Cob(II)yrinic acid a,c-diamide adenosyltransferase; Cobinamide/cobalamin adenosyltransferase; Corrinoid adenosyltransferase; methylmalonic aciduria (cobalamin deficiency) cblB type; methylmalonic aciduria (cobalamin deficiency) type B homolog (human); methylmalonic aciduria type B homolog; Methylmalonic aciduria type B protein; Mmab | |
Full length human recombinant protein of human FKBPL produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction