Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMAB Monoclonal Antibody (OTI5G1)
GREENER_CHOICE

Catalog No. PIMA525381
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525381 Supplier Invitrogen™ Supplier No. MA525381
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MMAB Monoclonal Antibody for Western Blot, IHC (P), Flow

This gene encodes a protein that catalyzes the final step in the conversion of vitamin B(12) into adenosylcobalamin (AdoCbl), a vitamin B12-containing coenzyme for methylmalonyl-CoA mutase. Mutations in the gene are the cause of vitamin B12-dependent methylmalonic aciduria linked to the cblB complementation group.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMAB
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5G1
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MMAB
Gene Accession No. Q96EY8
Gene Alias 9130222L19Rik; aquocob(I)alamin vitamin B12s adenosyltransferase; ATP:Cob(I)alamin Adenosyltransferase; ATP:corrinoid adenosyltransferase; ATR; cblB; CFAP23; cilia and flagella associated protein 23; cob; cob(I)alamin adenosyltransferase; cob(I)yrinic acid a,c-diamide adenosyltransferase, mitochondrial; Cob(II)alamin adenosyltransferase; Cob(II)yrinic acid a,c-diamide adenosyltransferase; Cobinamide/cobalamin adenosyltransferase; Corrinoid adenosyltransferase; methylmalonic aciduria (cobalamin deficiency) cblB type; methylmalonic aciduria (cobalamin deficiency) type B homolog (human); methylmalonic aciduria type B homolog; Methylmalonic aciduria type B protein; Mmab
Gene Symbols MMAB
Host Species Mouse
Immunogen Full length human recombinant protein of MMAB produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 326625
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.