Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMAB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528499
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528499 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528499 Supplier Invitrogen™ Supplier No. PA528499
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2. Predicted reactivity: Mouse (90%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein that catalyzes the final step in the conversion of vitamin B(12) into adenosylcobalamin (AdoCbl), a vitamin B12-containing coenzyme for methylmalonyl-CoA mutase. Mutations in the gene are the cause of vitamin B12-dependent methylmalonic aciduria linked to the cblB complementation group.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMAB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene MMAB
Gene Accession No. Q96EY8
Gene Alias 9130222L19Rik; aquocob(I)alamin vitamin B12s adenosyltransferase; ATP:Cob(I)alamin Adenosyltransferase; ATP:corrinoid adenosyltransferase; ATR; cblB; CFAP23; cilia and flagella associated protein 23; cob; cob(I)alamin adenosyltransferase; cob(I)yrinic acid a,c-diamide adenosyltransferase, mitochondrial; Cob(II)alamin adenosyltransferase; Cob(II)yrinic acid a,c-diamide adenosyltransferase; Cobinamide/cobalamin adenosyltransferase; Corrinoid adenosyltransferase; methylmalonic aciduria (cobalamin deficiency) cblB type; methylmalonic aciduria (cobalamin deficiency) type B homolog (human); methylmalonic aciduria type B homolog; Methylmalonic aciduria type B protein; Mmab
Gene Symbols MMAB
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MMAB. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 326625
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.