Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMACHC Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | MMACHC |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
MMACHC Polyclonal specifically detects MMACHC in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MMACHC | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
cblC, DKFZP564I122, FLJ25671, methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria, methylmalonic aciduria and homocystinuria type C protein, RP11-291L19.3 | |
MMACHC | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
25974 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:MEPKVAELKQKIEDTLCPFGFEVYPFQVAWYNELLPPAFHLPLPGPTLAFLVLSTPAMFDRALKPFLQSCHLRMLTDPVDQCVAYHLGRVRE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title