Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMACHC Antibody (OTI4E9), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272734JF549
Description
MMACHC Monoclonal specifically detects MMACHC in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
MMACHC | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MMACHC | |
Full length recombinant protein of human MMACHC (NP_056321) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry | |
OTI4E9 | |
Western Blot, Flow Cytometry | |
cblC, DKFZP564I122, FLJ25671, methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria, methylmalonic aciduria and homocystinuria type C protein, RP11-291L19.3 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
25974.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction