Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMACHC Mouse anti-Human, Clone: 4G10A11, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | MMACHC |
---|---|
Clone | 4G10A11 |
Concentration | 0.91 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7027
![]() ![]() |
Proteintech Group Inc
666091IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The exact function of the protein encoded by this gene is not known, however, its C-terminal region shows similarity to TonB, a bacterial protein involved in energy transduction for cobalamin (vitamin B12) uptake. Hence, it is postulated that this protein may have a role in the binding and intracellular trafficking of cobalamin. Mutations in this gene are associated with methylmalonic aciduria and homocystinuria type cblC.Specifications
MMACHC | |
0.91 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
cblC, MMACHC, RP11 291L19.3 | |
MMACHC | |
IgG1 | |
Protein G | |
Antibody |
4G10A11 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9Y4U1 | |
25974 | |
MMACHC Fusion Protein Ag15001 | |
Primary | |
-20°C | |
MMACHC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title