Learn More
MMACHC Mouse anti-Human, Clone: OTI6H5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504955

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The exact function of the MMACHC protein is not known, however, its C-terminal region shows similarity to TonB, a bacterial protein involved in energy transduction for cobalamin (vitamin B12) uptake. Hence, it is postulated that this protein may have a role in the binding and intracellular trafficking of cobalamin. Mutations in this gene are associated with methylmalonic aciduria and homocystinuria type cblC.Specifications
MMACHC | |
Monoclonal | |
Unconjugated | |
MMACHC | |
cblC | |
Mouse | |
Affinity Chromatography | |
RUO | |
25974 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI6H5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y4U1 | |
MMACHC | |
Full length human recombit protein of human MMACHC produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.