Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMACHC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587836
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587836 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587836 Supplier Invitrogen™ Supplier No. PA587836
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKPFLQSCHL RMLTDPVDQC VAYHLGRVRE SLPELQIEII ADYEVHPNRR PKILAQTAAH VAGAAYYYQR QDVEADPWGN QRISGVCIHP RFGGWFAIRG VVLLPGIEVP DLPPRKPHDC VPTRADRIAL LEGFNFHWRD WTYRDAVTPQ ERYSEEQKAY FSTPPAQRLA LLGLAQPSEK PSSPSPDLPF TTPAPKKPGN PSRARSWLSP RVSPPASPGP; Positive Samples: Raji, Jurkat, Mouse kidney, Rat brain; Cellular Location: Cytoplasm.

The exact function of the MMACHC protein is not known, however, its C-terminal region shows similarity to TonB, a bacterial protein involved in energy transduction for cobalamin (vitamin B12) uptake. Hence, it is postulated that this protein may have a role in the binding and intracellular trafficking of cobalamin. Mutations in this gene are associated with methylmalonic aciduria and homocystinuria type cblC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMACHC
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MMACHC
Gene Accession No. Q9CZD0, Q9Y4U1
Gene Alias 1810037K07Rik; cblC; Cyanocobalamin reductase (cyanide-eliminating); methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria; methylmalonic aciduria and homocystinuria type C protein; Methylmalonic aciduria and homocystinuria type C protein homolog; methylmalonic aciduria cblC type, with homocystinuria; Mmachc; RGD1310806; RP11-291L19.3
Gene Symbols MMACHC
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 63-282 of human MMACHC (NP_0563212).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25974, 313520, 67096
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.