Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMADHC Antibody (OTI1G4), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272739AF647
Description
MMADHC Monoclonal specifically detects MMADHC in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
MMADHC | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MMADHC | |
Recombinant protein fragment corresponding to amino acids 39-296 of human MMADHC (NP_056517) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI1G4 | |
Western Blot | |
cblD, chromosome 2 open reading frame 25, methylmalonic aciduria (cobalamin deficiency) cblD type, with homocystinuria, mitochondrial, protein C2orf25, mitochondrial | |
Mouse | |
Affinity Purified | |
metabolism | |
27249.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction