Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMADHC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557990
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557990 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557990 Supplier Invitrogen™ Supplier No. PA557990
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVLLEKFING AKEICYALRA EGYWADFIDP SSGLAFFGPY TNNTLFETDE RYRHLGFSVD DLGCCKVIRH SLWGTHVVVG SIFTNAT Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Involved in cobalamin metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMADHC
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MMADHC
Gene Accession No. Q9H3L0
Gene Alias 2010311D03Rik; AI314967; C2orf25; CblD; CL25022; HSPC161; likely ortholog of H. sapiens chromosome 2 open reading frame 25 (C2orf25); methylmalonic aciduria (cobalamin deficiency) cblD type, with homocystinuria; methylmalonic aciduria and homocystinuria type D homolog, mitochondrial; Methylmalonic aciduria and homocystinuria type D protein, mitochondrial; methylmalonic aciduria and homocystinuria, cblD type; Mmadhc; My011; protein C2orf25, mitochondrial
Gene Symbols MMADHC
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MMADHC. Recombinant protein control fragment (Product #RP-96189).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27249
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.