Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MMADHC Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548947
Description
Predicted to react with mouse and rat based on sequence homology.
Involved in cobalamin metabolism.
Specifications
MMADHC | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9H3L0 | |
MMADHC | |
KLH conjugated synthetic peptide between 226-253 amino acids from the C-terminal region of human MMADHC. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MMADHC | |
2010311D03Rik; AI314967; C2orf25; CblD; CL25022; HSPC161; likely ortholog of H. sapiens chromosome 2 open reading frame 25 (C2orf25); methylmalonic aciduria (cobalamin deficiency) cblD type, with homocystinuria; methylmalonic aciduria and homocystinuria type D homolog, mitochondrial; Methylmalonic aciduria and homocystinuria type D protein, mitochondrial; methylmalonic aciduria and homocystinuria, cblD type; Mmadhc; My011; protein C2orf25, mitochondrial | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
27249 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction