Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116278
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116278 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116278 Supplier Invitrogen™ Supplier No. PA5116278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MMD2 Polyclonal Antibody for Western Blot

This gene encodes a member of the PAQR (progestin and adipoQ receptor) family. Members of this family are evolutionarily conserved with significant sequence identity to bacterial hemolysin-like proteins and are defined by a set of seven transmembrane domains. The protein encoded by this gene localizes to the Golgi apparatus to modulate Ras signaling. Alternative splicing results in multiple transcript variants and protein isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMD2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MMD2
Gene Accession No. Q8IY49, Q8R189
Gene Alias 4930518M15Rik; C88001; Mmd2; monocyte to macrophage differentiation associated 2; monocyte to macrophage differentiation factor 2; monocyte to macrophage differentiation-associated 2; monocyte-to-macrophage differentiation factor 2; monocyte-to-macrophage differentiation-associated protein 2; PAQR10; Progestin and adipoQ receptor family member 10; progestin and adipoQ receptor family member X
Gene Symbols MMD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 6-35 amino acids from the N-terminal region of human MMD2.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100505381, 221938, 75104
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.