Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMD2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA544184

Catalog No. PIPA544184


Only null left
Add to Cart

Description

Description

Peptide sequence: APRLLDFQKT KYARFMNHRV PAHKRYQPTE YEHAANCATH AFWIIPSILG Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

This gene encodes a member of the PAQR (progestin and adipoQ receptor) family. Members of this family are evolutionarily conserved with significant sequence identity to bacterial hemolysin-like proteins and are defined by a set of seven transmembrane domains. The protein encoded by this gene localizes to the Golgi apparatus to modulate Ras signaling. Alternative splicing results in multiple transcript variants and protein isoforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MMD2
Polyclonal
Unconjugated
MMD2
4930518M15Rik; C88001; Mmd2; monocyte to macrophage differentiation associated 2; monocyte to macrophage differentiation factor 2; monocyte to macrophage differentiation-associated 2; monocyte-to-macrophage differentiation factor 2; monocyte-to-macrophage differentiation-associated protein 2; PAQR10; Progestin and adipoQ receptor family member 10; progestin and adipoQ receptor family member X
Rabbit
Affinity chromatography
RUO
100505381, 221938
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q8IY49
MMD2
Synthetic peptide directed towards the N-terminal of human MMD2 (aa 3-52).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.