Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MME Mouse anti-Human, Unconjugated, Clone: CB-CALLA, Abnova™

Catalog No. 89337801 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-801 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-801 Supplier Abnova Corporation Supplier No. MAB21310
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

MME Monoclonal antibody specifically detects MME in Human samples. It is validated for IF, Flow Cyt.

Specifications

Antigen MME
Applications Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone CB-CALLA
Conjugate Unconjugated
Description membrane metallo-endopeptidase
Gene MME
Gene Alias CALLA, CD10, DKFZp686O16152, MGC126681, MGC126707, NEP
Host Species Mouse
Immunogen Human CD10
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4311
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.