Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMEL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582339
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582339 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582339 Supplier Invitrogen™ Supplier No. PA582339
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVIEKRGSQP LLDILEVVGG WPVAMDRWNE TVGLEWELER QLALMNSQFN RRVLIDLFIW NDDQNSSRHI IYIDQPTLGM PSREYYFNGG SNRKVREAYL QFMVSVATLL REDANLPRDS CLVQEDMVQV LEL Highest antigen sequence indentity to the following orthologs: Mouse - 76%, Rat - 77%.

MMEL1 is a member of the neutral endopeptidase (NEP) or membrane metallo-endopeptidase (MME) family. Family members play important roles in pain perception, arterial pressure regulation, phosphate metabolism and homeostasis. MMEL1 is a type II transmembrane protein and is thought to be expressed as a secreted protein. The protein encoded by this gene is a member of the neutral endopeptidase (NEP) or membrane metallo-endopeptidase (MME) family. Family members play important roles in pain perception, arterial pressure regulation, phosphate metabolism and homeostasis. This protein is a type II transmembrane protein and is thought to be expressed as a secreted protein. This gene is expressed mainly in testis with weak expression in the brain, kidney, and heart.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMEL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MMEL1
Gene Accession No. Q495T6
Gene Alias EC 3.4.24; EC 3.4.24.11; mel transforming oncogene-like 1; Mell1; membrane metalloendopeptidase like 1; Membrane metallo-endopeptidase-like 1; Membrane metallo-endopeptidase-like 1, soluble form; Membrane metallo-endopeptidase-like 2; MGC119454; MGC119455; MGC119456; Mmel1; MMEL2; NEP2; NEP2(m); NEP2(s); NEPII; NEPIIMGC119456; NEPLP; NEPLP alpha; NEPLP beta; NEPLP gamma; neprilysin 2; neprilysin II; Neprilysin-2; Neprilysin-2 secreted; Neprilysin-like 1; neprilysin-like peptidase; Nl1; NL-1; NL2; NL2NL1; SEP; Soluble secreted endopeptidase; zinc metallopeptidase
Gene Symbols MMEL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MMEL1. Recombinant protein control fragment (Product #RP-88846).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79258
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.