Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mMESSAGE mMACHINE™ SP6 Transcription Kit

Catalog No. am1340
Encompass_Preferred
Click to view available options
Quantity:
25 Reactions

in vitro synthesis of large amounts of capped RNA

mMESSAGE mMACHINE™ kits are designed for in vitro synthesis of large amounts of capped RNA. Capped RNA mimics most eukaryotic mRNAs found in vivo, because it has a 7-methyl guanosine cap structure at the 5′ end.

  • RNA Yield: 5-25μg
  • Include cap analog [m7G(5′)ppp(5′)G] in an ultra high-yield transcription reaction
  • Cap analog is incorporated only as first or 5′ terminal G of transcript because its structure precludes its incorporation at other position in RNA molecule
  • Simplified reaction format in which all four ribonucleotides and cap analog are mixed in single solution
  • Ideal for routine synthesis of capped RNAs for oocyte microinjection, in vitro translation, transfection, and other applications

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage SP6 Enzyme Mix, 10X Reaction Buffer 2XNTP⁄CAP Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, and Gel Loading Buffer II are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Final Product Type Synthetic mRNA, mRNA
Format Kit
Sample Type mRNA, Synthetic mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions 25 Reactions
Product Line Ambion™, mMESSAGE mMACHINE™
Product Type SP6 In Vitro Transcription kit
Quantity 25 Reactions
Promoter SP6
Shipping Condition Dry Ice
Yield 15-25 μg
Starting Material PCR Products, Linearized DNA plasmid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.