Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MMG 11
GREENER_CHOICE

Catalog No. 685810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-581-0 10 mg
68-585-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 68-581-0 Supplier Tocris Bioscience™ Supplier No. 6858/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

TLR2 antagonist

MMG 11 is a TLR2 antagonist (IC50 values are in the range 1.7 - 5.7 μM). Exhibits selectivity for TLR2 over TLR4, TLR5, TLR7/8 and TLR9. Reduces TLR2-mediated cytokine secretion from PBMCs. MMG 11 preferentially inhibits TLR2/1- over TLR2/6-mediated NK-κB activation (IC50 values are 0.87 and 7.4 μM, respectively).

Specifications

Chemical Name or Material Ethyl 5-(2-oxo-2-(2,3,4-trihydroxyphenyl)ethyl)furan-2-carboxylate
CAS 313254-94-3
Quantity 10 mg
Target Toll-like Receptor Antagonists
Molecular Formula C15H14O7
Formula Weight 1414
Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.