Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMGT1 Monoclonal Antibody (OTI4D7)
GREENER_CHOICE

Catalog No. PIMA527396
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527396 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527396 Supplier Invitrogen™ Supplier No. MA527396
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MMGT1 Monoclonal Antibody for Western Blot, IHC (P)

Mmgt1 encodes a protein that that mediates Mg(2+) transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMGT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MMGT1
Gene Accession No. Q8N4V1
Gene Alias 9630048L06Rik; BC032271; EMBL:AAI68927.1}; EMC5; ER membrane protein complex subunit 5; Membrane magnesium transporter 1; membrane magnesium transporter 1 {ECO:0000250; Mir5116; mir-5116; Mmgt1; mmgt1 {ECO:0000250; RGD1566339; Tmem32; transmembrane protein 32; transmembrane protein 32 {ECO:0000312; UniProtKB:Q8K273}
Gene Symbols MMGT1
Host Species Mouse
Immunogen Full length human recombinant protein of MMGT1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93380
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.