Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMGT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597973
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597973 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597973 Supplier Invitrogen™ Supplier No. PA597973
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MMGT1 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Mmgt1 encodes a protein that that mediates Mg(2+) transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMGT1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MMGT1
Gene Accession No. Q8N4V1
Gene Alias 9630048L06Rik; BC032271; EMBL:AAI68927.1}; EMC5; ER membrane protein complex subunit 5; Membrane magnesium transporter 1; membrane magnesium transporter 1 {ECO:0000250; Mir5116; mir-5116; Mmgt1; mmgt1 {ECO:0000250; RGD1566339; Tmem32; transmembrane protein 32; transmembrane protein 32 {ECO:0000312; UniProtKB:Q8K273}
Gene Symbols MMGT1
Host Species Rabbit
Immunogen Recombinant Human Membrane magnesium transporter 1 protein (66-131AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93380
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.