Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ MMK 1
Click to view available options
Quantity:
1 mg
Description
MMK 1 is a potent and selective human formyl peptide receptor FPR2 agonist (EC50 values are 1, 2 and > 10 000 nM at mFRP2, hFPR2 and hFPR1 respectively). Induces migration of human monocytes and neutrophils via a chemotactic mechanism and enhances production of proinflammatory cytokines IL-1β and IL-6. Also activates the neutrophil superoxide-generating NADPH-oxidase.
DMSO is not recommended as a solvent for this peptide.
Specifications
Specifications
| Purity | 0.95 |
| Description | MMK 1 |
| Molecular Weight (g/mol) | 1610.97 |
| Product Type | MMK 1 |
| Quantity | 1 mg |
| Sequence | Leu-Glu-Ser-Ile-Phe-Arg-Ser-Leu-Leu-Phe-Arg-Val-Met |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction