Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-1 Antibody (6A5), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222123IR
Description
MMP-1 Monoclonal specifically detects MMP-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MMP-1 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MMP1 | |
Purified recombinant fragment of human MMP1 expressed in E. coli. [UniProt# P03956] | |
0.1 mL | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix | |
4312 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
6A5 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
CLGmatrix metalloprotease 1, CLGN, EC 3.4.24, EC 3.4.24.7, Fibroblast collagenase, interstitial collagenase, matrix metallopeptidase 1 (interstitial collagenase), matrix metalloproteinase 1 (interstitial collagenase), Matrix metalloproteinase-1, MMP-1 | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction