Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-10 Antibody (110316), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB9102K
Description
MMP-10 Monoclonal antibody specifically detects MMP-10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Flow Cytometry, CyTOFSpecifications
MMP-10 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation, Flow Cytometry, CyTOF | |
110316 | |
Western Blot, Immunohistochemistry, Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
EC 3.4.24, EC 3.4.24.22, matrix metallopeptidase 10 (stromelysin 2), matrix metalloprotease 10, matrix metalloproteinase 10 (stromelysin 2), Matrix metalloproteinase-10, MMP-10, SL-2, STMY2stromelysin-2, stromelysin 2, transin 2, transin-2 | |
Mouse myeloma cell line NS0-derived recombinant human MMP-10, Phe99-Cys476, Accession # P09238 | |
0.5 mL | |
Angiogenesis, Cancer, Extracellular Matrix | |
4319 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction