Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-10 Antibody (MM0024-9G10), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11060990PE
Description
MMP-10 Monoclonal antibody specifically detects MMP-10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| MMP-10 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0024-9G10 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 3.4.24, EC 3.4.24.22, matrix metallopeptidase 10 (stromelysin 2), matrix metalloprotease 10, matrix metalloproteinase 10 (stromelysin 2), Matrix metalloproteinase-10, MMP-10, SL-2, STMY2stromelysin-2, stromelysin 2, transin 2, transin-2 | |
| Human recombinant MMP-10 | |
| 0.1 mL | |
| Angiogenesis, Cancer, Extracellular Matrix | |
| 4319 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction