Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-10 (Stromelysin-2) Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS822P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS822P 1 mL 200μg/mL
MS822P0 100 μL 200μg/mL
MS822P1 500 μL 200μg/mL
MS822P1ABX 100 μL 1mg/mL
MS822PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS822P Supplier Epredia Supplier No. MS822P

Epredia™ MMP-10 (Stromelysin-2) Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Matrix metalloproteinases (MMP) are proteolytic enzymes capable of degrading extracellular matrix. Stromelysin, a member of the matrix metalloproteinase family, demonstrates wide substrate specificity with the ability to degrade proteoglycan, fibronectin, laminin, casein, and the nonhelical region of collagen. The two forms of stromelysin (SL), types 1 (MMP-3) and 2 (MMP-10), share 82% sequence homology, but exhibit differences in cellular synthesis and inducibility by cytokines and growth factors in vitro.

Host Species: Mouse

Clone: IVC5

Isotype: IgG2b

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human recombinant MMP-10 (stromelysin-2 protein)

Molecular Weight: ∽57kDa (proform) and ∽44kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) human large cell lymphoma U937 cells; Placenta; Bladder, breast and ovarian CAs

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen MMP-10 (Stromelysin-2) Ab-2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone IVC5
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human recombinant MMP-10 (stromelysin-2 protein)
Quantity 1 mL
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.