Learn More
Description
Host Species: Mouse
Clone: IVC5
Isotype: IgG2b
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Human recombinant MMP-10 (stromelysin-2 protein)
Molecular Weight: ∽57kDa (proform) and ∽44kDa (active form)
Positive Control: Conditioned, serum-free medium from (TPA-treated) human large cell lymphoma U937 cells; Placenta; Bladder, breast and ovarian CAs
Cellular Localization: Cytoplasmic
Recommended for:
- Western Blotting
- Immunoprecipitation (Native verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | MMP-10 (Stromelysin-2) Ab-2 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | IVC5 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human recombinant MMP-10 (stromelysin-2 protein) |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.