Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-12 Antibody (2G3), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | MMP-12 |
---|---|
Clone | 2G3 |
Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
MMP-12 Monoclonal antibody specifically detects MMP-12 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
MMP-12 | |
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
4321 | |
IgG | |
Affinity purified |
2G3 | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
EC 3.4.24, hME, HMEEC 3.4.24.65, Macrophage elastase, macrophage metalloelastase, matrix metallopeptidase 12 (macrophage elastase), matrix metalloproteinase 12 (macrophage elastase), Matrix metalloproteinase-12, ME, MGC138506, MME, MMP-12 | |
A synthesized peptide derived from Human MMP-12 [UniProt P39900] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title