Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-12 Antibody (701E4.03), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals DDX0284P100
Description
MMP-12 Monoclonal specifically detects MMP-12 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
MMP-12 | |
Monoclonal | |
0.5 mg/mL | |
ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
MMP12 | |
MMP-12/HME was used as immunogen. | |
0.1 mg | |
Primary | |
Human | |
IgG1 |
ELISA, Immunoprecipitation | |
701E4.03 | |
Unconjugated | |
EC 3.4.24, hME, HMEEC 3.4.24.65, Macrophage elastase, macrophage metalloelastase, matrix metallopeptidase 12 (macrophage elastase), matrix metalloproteinase 12 (macrophage elastase), Matrix metalloproteinase-12, ME, MGC138506, MME, MMP-12 | |
Mouse | |
Ion exchange chromatography | |
RUO | |
4321 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction