Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-13 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 3 publications

$243.50

Specifications

Antigen MMP-13
Concentration 0.5 mg/mL
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP145723SS
SDP
View Documents
Novus Biologicals
NBP145723SS
0.025 mg
Each for $243.50
Only null left
Add to Cart
 
NBP145723
SDP
View Documents
Novus Biologicals
NBP145723
0.1 mg N/A N/A N/A
Description

Description

MMP-13 Polyclonal specifically detects MMP-13 in Human, Mouse, Rat, Bovine, Equine, Rabbit samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

MMP-13
Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin
Polyclonal
Rabbit
Cancer, Cellular Markers, Extracellular Matrix
PBS (pH 7.2), 5 mM EDTA, 0.5% BSA, 30% Glycerol with 0.03% Proclin 300
CLG 3, CLG3EC 3.4.24, collagenase 3, EC 3.4.24.-, EC 3.4.24.22, EC 3.4.24.24, EC 3.4.24.35, EC 3.4.24.65, EC 3.4.24.7, MANDP1, matrix metallopeptidase 13 (collagenase 3), matrix metalloproteinase 13 (collagenase 3), Matrix metalloproteinase-13, MMP-13
MMP13
IgG
Affinity Purified
The antibody recognizes the proenzyme (∼60 kDa) and the cleaved fragments (34 kDa and 48 kDa) of MMP-13.
0.5 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat, Bovine, Equine, Rabbit
P23097
4322
Synthetic peptide surrounding amino acid 454 of rat MMP-13.
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.