Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-13 Antibody (OTI2D8), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272740V
Description
MMP-13 Monoclonal specifically detects MMP-13 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
MMP-13 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MMP13 | |
Recombinant protein fragment corresponding to amino acids 104-471 of human MMP-13 (NP_002418) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown | |
OTI2D8 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, KnockDown Validated | |
CLG 3, CLG3EC 3.4.24, collagenase 3, EC 3.4.24.-, EC 3.4.24.22, EC 3.4.24.24, EC 3.4.24.35, EC 3.4.24.65, EC 3.4.24.7, MANDP1, matrix metallopeptidase 13 (collagenase 3), matrix metalloproteinase 13 (collagenase 3), Matrix metalloproteinase-13, MMP-13 | |
Mouse | |
Affinity Purified | |
Cancer, Cellular Markers, Extracellular Matrix | |
4322.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction