Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-14/MT1-MMP Antibody (MM0027-9E10) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
| Antigen | MMP-14/MT1-MMP |
|---|---|
| Clone | MM0027-9E10 |
| Concentration | LYOPH |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017988
![]() |
Novus Biologicals
NB110609930.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB11060993
![]() |
Novus Biologicals
NB11060993 |
0.1 mg |
Each for $560.50
|
|
|||||
Description
MMP-14/MT1-MMP Monoclonal specifically detects MMP-14/MT1-MMP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MMP-14/MT1-MMP | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.4.24, EC 3.4.24.80, matrix metallopeptidase 14 (membrane-inserted), matrix metalloproteinase 14 (membrane-inserted), matrix metalloproteinase-14, membrane type 1 metalloprotease, Membrane-type matrix metalloproteinase 1, Membrane-type-1 matrix metalloproteinase, MMP-14, MMP-X1, MT1MMP, MT1-MMPMTMMP1, MT-MMP 1 | |
| MMP14 | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| MM0027-9E10 | |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Extracellular Matrix | |
| P50281 | |
| 4323 | |
| IgG1 | |
| Protein G purified | |
| This antibody recognizes human MMP-14 in western blot without cross reactivity with human MMP-1-13, and MMP14-15. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title