Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-15 Polyclonal Antibody, Abbomax

Catalog No. 502203736
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3736 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3736 Supplier Abbomax Inc Supplier No. 5004124
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Human liver Cellular location: Secreted, extracellular space, extracellular matrix.

MMP-15 Polyclonal antibody specifically detects MMP-15 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MMP-15
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Mmp15
Gene Accession No. O54732, P51511
Gene Alias AI503551; matrix metallo protease; matrix metallopeptidase 15; matrix metallopeptidase 15 (membrane-inserted); matrix metalloproteinase 15; matrix metalloproteinase 15 (membrane-inserted); matrix metalloproteinase-15; Membrane type 2-MMP; membrane-type matrix metalloproteinase 2; Membrane-type-2 matrix metalloproteinase; MMP; Mmp15; MMP-15; MMPs; MT2MMP; MT2-MMP; MT-MMP 2; MTMMP2; SMCP-2
Host Species Rabbit
Immunogen Matrix MMP-15
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17388, 4324
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.