Learn More
Description
Host Species: Mouse
Clone: A-Gel VC2
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Human APMA-activated native 72kDa MMP-2
Molecular Weight: 72kDa (proform) and ∽66kDa (active form)
Positive Control: Placenta; Bladder, breast and ovarian CAs
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (not qualified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | MMP-2 (72kDa Collagenase IV) Ab-4 |
| Classification | Monoclonal |
| Clone | A-Gel VC2 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human APMA-activated native 72kDa MMP-2 |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Research Discipline | Proteases And Inhibitors |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.