Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-2 Antibody (MMP2/1501), Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $487.50
Specifications
Antigen | MMP-2 |
---|---|
Clone | MMP2/1501 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, SDS-Page |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25340420
![]() |
Novus Biologicals
NBP25340420UG |
20 μg |
Each for $243.50
|
|
|||||
NB253404100
![]() |
Novus Biologicals
NBP253404100UG |
100 μg |
Each for $487.50
|
|
|||||
NB253404200
![]() |
Novus Biologicals
NBP253404200UG |
200 μg | N/A | N/A | N/A | ||||
Description
MMP-2 Monoclonal specifically detects MMP-2 in Human samples. It is validated for Western Blot, Protein Array.Specifications
MMP-2 | |
0.2 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page | |
Unconjugated | |
Mouse | |
Angiogenesis, Apoptosis, Cancer, Cell Biology, Cytoskeleton Markers, Extracellular Matrix, HIF Target Genes, Hypoxia, Metastasis, Tumor Suppressors | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
4313 | |
Recombinant human MMP2 protein fragment (aa444-575) (exact sequence is proprietary) | |
Primary | |
Store at 4C. |
MMP2/1501 | |
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, SDS-Page | |
Monoclonal | |
Purified | |
RUO | |
Human | |
72 kDa gelatinase, CLG4, CLG4A72 kDa type IV collagenase, collagenase type IV-A, EC 3.4.24, EC 3.4.24.24, Gelatinase A, matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase), matrix metalloproteinase 2 (gelatinase A, 72kD gelatinase, 72kD type IVcollagenase), Matrix metalloproteinase-2, matrix metalloproteinase-II, MMP-2, MMP-II, MONA, neutrophil gelatinase, TBE-1matrix metalloproteinase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase) | |
MMP2 | |
IgG1 | |
Protein A or G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title